missing translation for 'onlineSavingsMsg'
Learn More

Dacarbazine 98.0+%, TCI America™

Catalog No. p-7127836
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36341G 1 g
D36345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36341G Supplier TCI America Supplier No. D36341G
Only null left

Chemical Identifiers

CAS 3-4-4342
Molecular Formula C6H10N6O
Molecular Weight (g/mol) 182.19
MDL Number MFCD00057167
InChI Key OMJKFYKNWZZKTK-UXBLZVDNSA-N
Synonym dacarbazine, biocarbazine, deticene, dtic-dome, biocarbazine r, decarbazine, dtic, icdt, dacarbazinum inn-latin, dacarbazino inn-spanish
PubChem CID 5353562
IUPAC Name (5E)-5-(dimethylaminohydrazinylidene)imidazole-4-carboxamide
SMILES CN(C)NN=C1C(=NC=N1)C(=O)N

Specifications

Melting Point 255°C
Color Yellow
Quantity 1 g
Formula Weight 182.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dacarbazine
RTECSNumber NI3950000
TSCA No
Recommended Storage Refrigerator