missing translation for 'onlineSavingsMsg'
Learn More

Dabsyl Chloride 98.0+%, TCI America™

Catalog No. p-7126701
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13821G 1 g
D138225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D13821G Supplier TCI America Supplier No. D13821G
Only null left

Chemical Identifiers

CAS 56512-49-3
Molecular Formula C14H14ClN3O2S
Molecular Weight (g/mol) 323.80
MDL Number MFCD00007444
InChI Key VTVWTPGLLAELLI-UHFFFAOYSA-N
Synonym dabsyl chloride, 4-dimethylaminoazobenzene-4'-sulfonyl chloride, 4-dimethylamino azobenzene-4'-sulfonyl chloride, p-dimethylaminoazobenzene-4-sulfonyl chloride, p-p-dimethylamino phenyl azo benzenesulfonyl chloride, benzenesulfonyl chloride, 4-4-dimethylamino phenyl azo, benzenesulfonyl chloride, p-p-dimethylamino phenyl azo, 4-n,n-dimethylaminoazobenzene-4'-sulfonyl chloride, p-4-dimethylamino phenyl azo benzenesulphonyl chloride
PubChem CID 91660
IUPAC Name 4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzene-1-sulfonyl chloride
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Quantity 1 g
UN Number 3261
Formula Weight 323.80
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dabsyl Chloride [N-Protecting Agent for Peptides Research]
RTECSNumber DB8927800
TSCA No