missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Dabco™ 33-LV

Catalog No. 290734100ML
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
290734100ML 100 mL
290734500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 290734100ML Supplier Sigma Aldrich Supplier No. 290734100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dabco(R)33-LV (Db) is a gelling catalyst and a bidentate ligand that forms a self-assembled monolayer (SAM) on a variety of substrates. It functionalizes the surface and immobilizes the surface atoms.Db can be used in the preparation of epoxy based soybean oil for the formation of polyurethane foams.DABCO is a registered trademark of Air Products and Chemicals, Inc.

Specifications

CAS 280-57-9
Density 1.02 g/mL
Molecular Formula C6H12N2
Linear Formula C6H12N2
Quantity 100 mL
MDL Number MFCD00006689
Synonym 1,4-Diazabicyclo[2.2.2 ]octane solution
Molecular Weight (g/mol) 112.17