Vector Laboratories DAB Enhancing Solution SK-4103-400
Catalog No. 502045702
DAB Enhancing Solution, 30 ml, intensifies the DAB reaction product in stained sections. A ten-second exposure to this solution darkens the reaction product.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
DAB Enhancing Solution, 30 ml, intensifies the DAB reaction product in stained sections. A ten-second exposure to this solution darkens the reaction product.