missing translation for 'onlineSavingsMsg'
Learn More

STERIS™ DA-7645 Detergent

Catalog No. 17018312
Change view
Click to view available options
Size:
5 gal.
55 gal.
Container Type:
Drum
Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Container Type
17018312 5 gal. Pail
17018313 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17018312 Supplier STERIS™ Supplier No. 116505

DA-7645, a proprietary blend of surfactants and solvents, enhances manual cleaning in pharmaceutical, biotech, and cosmetic industries. Its mild pH ensures operator safety, while comprehensive validation support makes it easy to use.

DA-7645 Process and Research Cleaner is specially formulated to enhance manual cleaning with enhanced substrate compatibility in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.

HOW DA-7645 WORKS

DA-7645 Detergent is a proprietary blend of surfactants, solvents and other performance-enhancing ingredients that provide multiple cleaning mechanisms in manual cleaning applications.

WHY DA-7645?

  • Efficacy - Mild pH formulation delivers excellent cleaning characteristics and substrate compatibility with enhanced operator safety.
  • Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports.
  • Productivity - Delivers efficiency to manual cleaning operations using a blend of surfactants, solvents, and other performance-enhancing ingredients.
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility.

Specifications

Product Type Detergent
Color-coded No
Form Clear Liquid
pH 8.9 (Diluted)
Scent Unscented
Sterility Non-sterile
Container Type Pail
Size 5 gal.