Learn More
Abcam Inc D4476, CK1 inhibitor, 50MG
Catalog No. 502707780
Encompass
MW 398.4 Da, Purity >99%. Selective cell-permeable inhibitor of casein kinase 1 (CK1) . (IC values are 0.3 (CK1), 0.5 (ALK5), 9.1 (PKd1) and 5.8 M (p38MAPK)). Displays selectivity over many other protein kinases.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502707780 Supplier Abcam Inc Supplier No. AB12022050MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 398.4 Da, Purity >99%. Selective cell-permeable inhibitor of casein kinase 1 (CK1) . (IC values are 0.3 (CK1), 0.5 (ALK5), 9.1 (PKd1) and 5.8 M (p38MAPK)). Displays selectivity over many other protein kinases.
The product is subject to the following: Abcam Restricted Use Statement