missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Xylose 98.0+%, TCI America™

Catalog No. p-7137957
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
X001925G 25 g
X0019500G 500 g
2 options

Catalog No. X001925G

Supplier: TCI America X001925G

Only null left

Chemical Identifiers

CAS 58-86-6
Molecular Formula C5H10O5
MDL Number MFCD00151475

Specifications

Melting Point 151°C
Color White
Quantity 25 g
Formula Weight 150.13
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Xylose
EINECSNumber (9)-0318
RTECSNumber ZF2285000
TSCA Yes