missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Xylose 98.0+%, TCI America™

Catalog No. X0019500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X0019500G 500 g
X001925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X0019500G Supplier TCI America Supplier No. X0019500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 58-86-6
Molecular Formula C5H10O5
MDL Number MFCD00151475

Specifications

Melting Point 151°C
Color White
Quantity 500 g
Formula Weight 150.13
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Xylose
EINECSNumber (9)-0318
RTECSNumber ZF2285000
TSCA Yes