missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Xylose, 99+%

Catalog No. AC141005000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC141005000 500 g Plastic bottle
AC141001000 100 g Plastic bottle
AC141000025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC141005000 Supplier Thermo Scientific Chemicals Supplier No. 141005000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 58-86-6 | C5H10O5 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58-86-6
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13

Specifications

Melting Point 148°C to 153°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 31,47
Merck Index 15,10285
Specific Rotation Condition + 20.00 (20.00°C c=10,H2O)
Specific Rotation + 20
Solubility Information Solubility in water: soluble. Other solubilities: soluble in pyridine and hot alcohol
Formula Weight 150.13
Percent Purity 99+%
Physical Form Fine Crystalline Powder
Chemical Name or Material D(+)-Xylose
Show More Show Less
EINECSNumber 200-400-7

RUO – Research Use Only