missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(+)-Xylose, 98+%

Catalog No. p-7080908
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1064322 100 g
AAA1064336 500 g
AAA106430E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1064322 Supplier Thermo Scientific Chemicals Supplier No. A1064322
Only null left

CAS: 58-86-6 | C5H10O5

D-(+)-Xylose is used to prepare furfural by acid catalyzed degradation reaction, which acts as a solvent as well as a precursor to synthetic polymers. It is a useful animal medicine involved in the treatment of malabsorption. It finds application in the production of sugar substitute xylitol by subjecting xylose to calyticl hydrogenation process. It is found that it undergoes an atmospheric-pressure enzyme (polyphosphate xylulokinase) driven process to get hydrogen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-Xylose is used to prepare furfural by acid catalyzed degradation reaction, which acts as a solvent as well as a precursor to synthetic polymers. It is a useful animal medicine involved in the treatment of malabsorption. It finds application in the production of sugar substitute xylitol by subjecting xylose to calyticl hydrogenation process. It is found that it undergoes an atmospheric-pressure enzyme (polyphosphate xylulokinase) driven process to get hydrogen.

Solubility
Soluble in water.

Notes
Store in cool place. Hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 58-86-6
Molecular Formula C5H10O5
MDL Number MFCD00151475

Specifications

Melting Point 147°C to 151°C
Density 1.525
Flash Point > 100°C (212°F)
Assay Percent Range >98%
Quantity 100 g
Beilstein 1562108
Sensitivity Hygroscopic
Merck Index 14,10087
Solubility Information Soluble in water.
Optical Rotation +19° (c=10 in water + NH4OH)
Formula Weight 150.13
Percent Purity ≥98%
Chemical Name or Material D- (+)-Xylose
Show More Show Less
EINECSNumber 200-400-7
RTECSNumber ZF2285000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only