missing translation for 'onlineSavingsMsg'
Learn More

d-Vitamin E Succinate, USP, 96-102%, Spectrum™ Chemical

Catalog No. 18602359
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602359 25 g Amber Glass Bottle
18602360 100 g Amber Glass Bottle
18602361 500 g Amber Glass Bottle
18602362 1 kg Amber Glass Bottle
18602363 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18602359 Supplier Spectrum Chemical Mfg Cor Supplier No. V103525GM
Get Up to $500 in Chemicals with Purchase
View all promotions

V1035, 4345-03-3, C33H54O5

Spectrum™ Chemical d-Vitamin E Succinate, USP is a fat-soluble antioxidant. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 4345-03-3
Molecular Formula C33H54O5
Molecular Weight (g/mol) 530.79
InChI Key IELOKBJPULMYRW-NJQVLOCASA-N
IUPAC Name 4-oxo-4-{[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl]oxy}butanoic acid
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(OC(=O)CCC(O)=O)=C(C)C(C)=C2O1

Specifications

CAS Max % 100%
Melting Point 75°C
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 96 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material RRR-Vitamin E Succinate