missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Valinol 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 103.17 |
| Percent Purity | ≥98.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | D-Valinol |
Safety and Handling
| TSCA | No |