Learn More
D-Valine tert-butyl ester hydrochloride, 95%
CAS: 104944-18-5 | C9H20ClNO2 | 209.71 g/mol
Supplier: Thermo Scientific Chemicals H6213114
Description
D-Valine tert-butyl ester hydrochloride used as a intermediate in pharmaceutical. Used as a an antitumor drug.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-Valine tert-butyl ester hydrochloride used as a intermediate in pharmaceutical. Used as a an antitumor drug.
Notes
Store at -20°C. Keep container tightly closed.
Specifications
| D-Valine tert-butyl ester hydrochloride | |
| C9H20ClNO2 | |
| h-d-val-otbu.hcl, d-valine tert-butyl ester hydrochloride, h-d-val-otbu hcl, r-tert-butyl 2-amino-3-methylbutanoate hydrochloride, tert-butyl 2r-2-amino-3-methylbutanoate hydrochloride, d-valine, 1,1-dimethylethyl ester, hydrochloride, d-valine tert.butyl ester hydrochloride, d-valine tert.butyl ester hcl, h-d-val-otbucl, h-d-val-otbu. hcl | |
| Cl.CC(C)[C@@H](N)C(=O)OC(C)(C)C | |
| 25 g | |
| 45108216 | |
| 95% |
| 104944-18-5 | |
| MFCD00237308 | |
| AUIVQIHTTVPKFS-OGFXRTJISA-N | |
| tert-butyl 2-amino-3-methylbutanoate hydrochloride | |
| 209.71 | |
| 209.71 |
Chemical Identifiers
| 104944-18-5 | |
| 209.71 | |
| AUIVQIHTTVPKFS-OGFXRTJISA-N | |
| 45108216 |
| C9H20ClNO2 | |
| MFCD00237308 | |
| h-d-val-otbu.hcl, d-valine tert-butyl ester hydrochloride, h-d-val-otbu hcl, r-tert-butyl 2-amino-3-methylbutanoate hydrochloride, tert-butyl 2r-2-amino-3-methylbutanoate hydrochloride, d-valine, 1,1-dimethylethyl ester, hydrochloride, d-valine tert.butyl ester hydrochloride, d-valine tert.butyl ester hcl, h-d-val-otbucl, h-d-val-otbu. hcl | |
| Cl.CC(C)[C@@H](N)C(=O)OC(C)(C)C |
Safety and Handling
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only