missing translation for 'onlineSavingsMsg'
Learn More

D-Valine tert-butyl ester hydrochloride, 95%

Catalog No. AAH6213103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6213103 1 g
AAH6213106 5 g
AAH6213114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6213103 Supplier Thermo Scientific Chemicals Supplier No. H6213103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 104944-18-5 | C9H20ClNO2 | 209.71 g/mol

D-Valine tert-butyl ester hydrochloride used as a intermediate in pharmaceutical. Used as a an antitumor drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Valine tert-butyl ester hydrochloride used as a intermediate in pharmaceutical. Used as a an antitumor drug.

Notes
Store at -20°C. Keep container tightly closed.

Chemical Identifiers

CAS 104944-18-5
Molecular Formula C9H20ClNO2
Molecular Weight (g/mol) 209.71
MDL Number MFCD00237308
InChI Key AUIVQIHTTVPKFS-OGFXRTJISA-N
Synonym h-d-val-otbu.hcl, d-valine tert-butyl ester hydrochloride, h-d-val-otbu hcl, r-tert-butyl 2-amino-3-methylbutanoate hydrochloride, tert-butyl 2r-2-amino-3-methylbutanoate hydrochloride, d-valine, 1,1-dimethylethyl ester, hydrochloride, d-valine tert.butyl ester hydrochloride, d-valine tert.butyl ester hcl, h-d-val-otbucl, h-d-val-otbu. hcl
PubChem CID 45108216
SMILES Cl.CC(C)[C@@H](N)C(=O)OC(C)(C)C

Specifications

IUPAC Name tert-butyl (2R)-2-amino-3-methylbutanoate;hydrochloride
Quantity 1 g
Formula Weight 209.71
Percent Purity 95%
Chemical Name or Material D-Valine tert-butyl ester hydrochloride
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only