missing translation for 'onlineSavingsMsg'
Learn More

D-Tyrosine tert-butyl ester, 98%

CAS: 87553-74-0 | C13H19NO3 | 237.30 g/mol

$64.07 - $212.40

Chemical Identifiers

CAS 87553-74-0
Molecular Formula C13H19NO3
Molecular Weight (g/mol) 237.30
MDL Number MFCD00191035
InChI Key DIGHFXIWRPMGSA-LDGXTIHJNA-N
Synonym h-d-tyr-otbu, tert-butyl 2r-2-amino-3-4-hydroxyphenyl propanoate, d-tyrosine,1,1-dimethylethyl ester, d-tyrosine tert-butyl ester, d-tyrosine tert.butyl ester, d-tyrosine-t-butylester, r-tert-butyl 2-amino-3-4-hydroxyphenyl propanoate, r-2-amino-3-4-hydroxy-phenyl-propionic acid tert-butyl ester
PubChem CID 7408333
IUPAC Name tert-butyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoate
SMILES CC(C)(C)OC(=O)[C@H](N)CC1=CC=C(O)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6327403
View Documents
Thermo Scientific Chemicals
H6327403
1 g
Each for $64.07
Only null left
 
AAH6327406
View Documents
Thermo Scientific Chemicals
H6327406
5 g
Each for $212.40
Only null left
 
Description

Description

D-Tyrosine tert-butyl ester, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Tyrosine tert-butyl ester, is used as a pharmaceutical intermediate.

Solubility
Very slightly soluble in cold water.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

87553-74-0
237.30
DIGHFXIWRPMGSA-LDGXTIHJNA-N
7408333
CC(C)(C)OC(=O)[C@H](N)CC1=CC=C(O)C=C1
C13H19NO3
MFCD00191035
h-d-tyr-otbu, tert-butyl 2r-2-amino-3-4-hydroxyphenyl propanoate, d-tyrosine,1,1-dimethylethyl ester, d-tyrosine tert-butyl ester, d-tyrosine tert.butyl ester, d-tyrosine-t-butylester, r-tert-butyl 2-amino-3-4-hydroxyphenyl propanoate, r-2-amino-3-4-hydroxy-phenyl-propionic acid tert-butyl ester
tert-butyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoate

Specifications

87553-74-0
MFCD00191035
Very slightly soluble in cold water.
CC(C)(C)OC(=O)[C@H](N)CC1=CC=C(O)C=C1
1 g
7408333
98%
D-Tyrosine tert-butyl ester
C13H19NO3
h-d-tyr-otbu, tert-butyl 2r-2-amino-3-4-hydroxyphenyl propanoate, d-tyrosine,1,1-dimethylethyl ester, d-tyrosine tert-butyl ester, d-tyrosine tert.butyl ester, d-tyrosine-t-butylester, r-tert-butyl 2-amino-3-4-hydroxyphenyl propanoate, r-2-amino-3-4-hydroxy-phenyl-propionic acid tert-butyl ester
DIGHFXIWRPMGSA-LDGXTIHJNA-N
tert-butyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoate
237.30
237.14
Solid
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only