missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Tryptophan 98.0+%, TCI America™
$48.13 - $182.89
Chemical Identifiers
| CAS | 153-94-6 |
|---|---|
| Molecular Formula | C11H12N2O2 |
| Molecular Weight (g/mol) | 204.23 |
| MDL Number | MFCD00005647 |
| InChI Key | QIVBCDIJIAJPQS-SECBINFHSA-N |
| Synonym | d-tryptophan, h-d-trp-oh, r-tryptophan, d +-tryptophan, d-trytophane, d-tryptophane, tryptophan, d, 2r-2-amino-3-1h-indol-3-yl propanoic acid, +-tryptophan, h-d-typ-oh |
| PubChem CID | 9060 |
| ChEBI | CHEBI:16296 |
| IUPAC Name | (2R)-2-amino-3-(1H-indol-3-yl)propanoic acid |
| SMILES | N[C@H](CC1=CNC2=CC=CC=C12)C(O)=O |
Chemical Identifiers
| 153-94-6 | |
| 204.23 | |
| QIVBCDIJIAJPQS-SECBINFHSA-N | |
| 9060 | |
| (2R)-2-amino-3-(1H-indol-3-yl)propanoic acid |
| C11H12N2O2 | |
| MFCD00005647 | |
| d-tryptophan, h-d-trp-oh, r-tryptophan, d +-tryptophan, d-trytophane, d-tryptophane, tryptophan, d, 2r-2-amino-3-1h-indol-3-yl propanoic acid, +-tryptophan, h-d-typ-oh | |
| CHEBI:16296 | |
| N[C@H](CC1=CNC2=CC=CC=C12)C(O)=O |
Specifications
| 153-94-6 | |
| C11H12N2O2 | |
| 5 g | |
| QIVBCDIJIAJPQS-SECBINFHSA-N | |
| (2R)-2-amino-3-(1H-indol-3-yl)propanoic acid | |
| 9060 | |
| 204.23 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00005647 | |
| d-tryptophan, h-d-trp-oh, r-tryptophan, d +-tryptophan, d-trytophane, d-tryptophane, tryptophan, d, 2r-2-amino-3-1h-indol-3-yl propanoic acid, +-tryptophan, h-d-typ-oh | |
| N[C@H](CC1=CNC2=CC=CC=C12)C(O)=O | |
| 204.23 | |
| CHEBI:16296 | |
| ≥98.0% (HPLC,T) | |
| D-Tryptophan |
Safety and Handling
RTECSNumber : YN6129000
TSCA : Yes