missing translation for 'onlineSavingsMsg'
Learn More

D-Tryptophan Methyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. p-7137218
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24425G 5 g
T244225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24425G Supplier TCI America Supplier No. T24425G
Only null left

Chemical Identifiers

CAS 14907-27-8
Molecular Formula C12H15ClN2O2
Molecular Weight (g/mol) 254.71
MDL Number MFCD00038992
InChI Key XNFNGGQRDXFYMM-UHFFFAOYNA-N
Synonym d-tryptophan methyl ester hydrochloride, h-d-trp-ome.hcl, r-methyl 2-amino-3-1h-indol-3-yl propanoate hydrochloride, methyl 2r-2-amino-3-1h-indol-3-yl propanoate hydrochloride, methyl d-tryptophanate hydrochloride, d-tryptophan methyl ester hcl, h-d-trp-ome hcl, dtrp-ome.hcl, pubchem10879, ksc174m8r
PubChem CID 11448200
IUPAC Name methyl 2-amino-3-(1H-indol-3-yl)propanoate hydrochloride
SMILES Cl.COC(=O)C(N)CC1=CNC2=CC=CC=C12

Specifications

Melting Point 205°C
Quantity 5 g
Formula Weight 254.71
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material D-Tryptophan Methyl Ester Hydrochloride
EINECSNumber (5)-6818
TSCA No