missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Tryptophan methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Description
D-Tryptophan Methyl Ester Hydrochloride is an intermediate in the synthesis of Tryptophan derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-Tryptophan Methyl Ester Hydrochloride is an intermediate in the synthesis of Tryptophan derivatives.
Solubility
Soluble in water (150 mg/ml), dimethyl sulfoxide and methanol (50 mg/ml).
Notes
Incompatible with strong oxidizing agents. Store under room temperature.
D-Tryptophan Methyl Ester Hydrochloride is an intermediate in the synthesis of Tryptophan derivatives.
Solubility
Soluble in water (150 mg/ml), dimethyl sulfoxide and methanol (50 mg/ml).
Notes
Incompatible with strong oxidizing agents. Store under room temperature.
Specifications
Specifications
| Melting Point | 213°C to 216°C |
| Solubility Information | Soluble in water (150mg/ml),dimethyl sulfoxide and methanol (50mg/ml). |
| Quantity | 1 g |
| Formula Weight | 254.71 |
| Optical Rotation | −18° (c=5 in Methanol) |
| Percent Purity | 98% |
| Chemical Name or Material | D-Tryptophan methyl ester hydrochloride |
Safety and Handling
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only