missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Tryptophan, 99%

Catalog No. p-7024163
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1842606 5 g
AAA1842614 25 g
AAA1842622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1842606 Supplier Thermo Scientific Chemicals Supplier No. A1842606
Only null left

CAS: 153-94-6 | C11H12N2O2 | 204.23 g/mol

D-Tryptophan is used for human embryonic kidney cell (HEK-293, ATCC: CRL-1573) culture. The product is a sweetener used to study the release of incretins from enteroendocrine cells triggered by sugar but not by sweeteners.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Tryptophan is used for human embryonic kidney cell (HEK-293, ATCC: CRL-1573) culture. The product is a sweetener used to study the release of incretins from enteroendocrine cells triggered by sugar but not by sweeteners.

Solubility
Soluble in 0.5M hydrochloric acid (50 mg/ml), water (11 mg/ml at 20°C), hot alcohol, and dimethyl sulfoxide. Insoluble in chloroform.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 153-94-6
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.23
MDL Number MFCD00005647
InChI Key QIVBCDIJIAJPQS-SECBINFHSA-N
Synonym d-tryptophan, h-d-trp-oh, r-tryptophan, d +-tryptophan, d-trytophane, d-tryptophane, tryptophan, d, 2r-2-amino-3-1h-indol-3-yl propanoic acid, +-tryptophan, h-d-typ-oh
PubChem CID 9060
ChEBI CHEBI:16296
IUPAC Name (2R)-2-amino-3-(1H-indol-3-yl)propanoic acid
SMILES N[C@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Melting Point 273°C (decomposition)
Beilstein 86198
Solubility Information Soluble in 0.5M hydrochloric acid (50mg/ml),water (11mg/ml at 20°C),hot alcohol,and dimethyl sulfoxide. Insoluble in chloroform.
Quantity 5 g
Formula Weight 204.23
Optical Rotation +31.5° (c=1 in Water)
Percent Purity 99%
Chemical Name or Material D-Tryptophan
EINECSNumber 205-819-9
RTECSNumber YN6129000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only