missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Trehalose Dihydrate, MP Biomedicals™
Used as a cryoprotectant in a variety of cell freezing media. D-(+)-Trehalose Dihydrate, MP Biomedicals is a non-reducing sugar formed from two glucose units joined by a 1-1 alpha bond. It is thought to provide plants and animals with the ability to withs
$2,928.83
Chemical Identifiers
| CAS | 6138-23-4 |
|---|---|
| Molecular Formula | C12H26O13 |
| Molecular Weight (g/mol) | 378.33 |
| MDL Number | MFCD00071594 |
| InChI Key | DPVHGFAJLZWDOC-DJCWUSJTNA-N |
| Synonym | trehalose dihydrate, d-+-trehalose dihydrate, a,a-trehalose, d +-trehalose dihydrate, unii-7yin7j07x4, alpha,alpha-trehalose dihydrate, d-trehalose dihydrate, mycose,1-o-alpha-d-glucopyranosyl-alpha-d-glucopyranoside, alp,alp.-trehalose, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran-3,4,5-triol |
| PubChem CID | 181978 |
| IUPAC Name | (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate |
| SMILES | O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MP215394891
|
N/A | 1 kg |
Each for $2,928.83
|
|
|||||
Description
Trehalose is a natural alpha-linked, non-reducing disaccharide formed by an α,α-1,1-glucoside bond between two α-glucose units, which yields 2 moles of D-glucose upon acid hydrolysis.
- Has 90% of the calorific content of sucrose
- Inhibits polyglutamine-mediated protein aggregation both in vivo and in vitro
- Oral administration of trehalose has been shown to effectively extend lifespan in a murine Huntington disease model
- Known to accumulate in osmotically-stressed cultured bacterial cells
- Protects against the adverse effects of dehydration
Chemical Identifiers
| 6138-23-4 | |
| 378.33 | |
| DPVHGFAJLZWDOC-DJCWUSJTNA-N | |
| 181978 | |
| O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O |
| C12H26O13 | |
| MFCD00071594 | |
| trehalose dihydrate, d-+-trehalose dihydrate, a,a-trehalose, d +-trehalose dihydrate, unii-7yin7j07x4, alpha,alpha-trehalose dihydrate, d-trehalose dihydrate, mycose,1-o-alpha-d-glucopyranosyl-alpha-d-glucopyranoside, alp,alp.-trehalose, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran-3,4,5-triol | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate |
Specifications
| 6138-23-4 | |
| MFCD00071594 | |
| Soluble in water (50mg/mL, clear, colorless solution) or hot alcohol. | |
| O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O | |
| 378.33 | |
| 378.3 | |
| Powder |
| C12H26O13 | |
| trehalose dihydrate, d-+-trehalose dihydrate, a,a-trehalose, d +-trehalose dihydrate, unii-7yin7j07x4, alpha,alpha-trehalose dihydrate, d-trehalose dihydrate, mycose,1-o-alpha-d-glucopyranosyl-alpha-d-glucopyranoside, alp,alp.-trehalose, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran-3,4,5-triol | |
| DPVHGFAJLZWDOC-DJCWUSJTNA-N | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate | |
| 181978 | |
| ≥99% |
Safety and Handling
Recommended Storage : Store at Room Temperature (15-30°C).