missing translation for 'onlineSavingsMsg'
Learn More

D-δ-Tocopherol, 93%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 421055000

 View more versions of this product

Catalog No. AC421055000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

D-δ-Tocopherol
>93°C
91% min. (GC)
C27H46O2
500mg
delta-tocopherol, unii-r0zb2556p8 component, unii-zl0rfa4e5n component, s-2,8-dimethyl-2-4s,8s-4,8,12-trimethyltridecyl-3,4-dihydro-2h-1-benzopyran-6-ol, 2s-2,8-dimethyl-2-4s,8s-4,8,12-trimethyltridecyl-3,4-dihydro-1-benzopyran-6-ol, 2h-1-benzopyran-6-ol,3,4-dihydro-2,8-dimethyl-2-4r,8r-4,8,12-trimethyltridecyl-, 2r
GZIFEOYASATJEH-VHFRWLAGSA-N
(2R)-2,8-dimethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol
12444418
93%
119-13-1
Authentic
Glass bottle
MFCD20486794
15,9656
Solubility in water: insoluble. Other solubilities: soluble in ethanol, acetone, chloroform, ether,, oils, fats
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=CC(O)=CC(C)=C2O1
402.66
402.65

Chemical Identifiers

119-13-1
402.66
GZIFEOYASATJEH-VHFRWLAGSA-N
12444418
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=CC(O)=CC(C)=C2O1
C27H46O2
MFCD20486794
delta-tocopherol, unii-r0zb2556p8 component, unii-zl0rfa4e5n component, s-2,8-dimethyl-2-4s,8s-4,8,12-trimethyltridecyl-3,4-dihydro-2h-1-benzopyran-6-ol, 2s-2,8-dimethyl-2-4s,8s-4,8,12-trimethyltridecyl-3,4-dihydro-1-benzopyran-6-ol, 2h-1-benzopyran-6-ol,3,4-dihydro-2,8-dimethyl-2-4r,8r-4,8,12-trimethyltridecyl-, 2r
(2R)-2,8-dimethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-299-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only