missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Sucrose 99.0+%, TCI America™

Catalog No. p-7135817
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S011125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S011125G Supplier TCI America Supplier No. S011125G
Only null left

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym D-(+)-Saccharose
PubChem CID 5988
ChEBI CHEBI:17992
IUPAC Name (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 187°C
Formula Weight 342.30
Percent Purity ≥99.0% (HPLC)
Color White
Quantity 25 g
Physical Form Crystal/Powder
Chemical Name or Material D-(+)-Sucrose
RTECSNumber WN6500000
TSCA Yes