missing translation for 'onlineSavingsMsg'
Learn More

D-Sphingosine 96.0+%, TCI America™

Catalog No. S087425MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S087425MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S087425MG Supplier TCI America Supplier No. S087425MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123-78-4
Molecular Formula C18H37NO2
Molecular Weight (g/mol) 299.499
MDL Number MFCD00036751
InChI Key WWUZIQQURGPMPG-KRWOKUGFSA-N
Synonym sphingosine, d-erythro-sphingosine, 4-sphingenine, d-sphingosine, sphing-4-enine, cerebroside, sphingenine, 4-trans-sphingenine, trans-d-erythro-2-amino-4-octadecene-1,3-diol, 2s,3r,4e-2-amino-4-octadecene-1,3-diol
PubChem CID 5280335
ChEBI CHEBI:16393
IUPAC Name (E,2S,3R)-2-aminooctadec-4-ene-1,3-diol
SMILES CCCCCCCCCCCCCC=CC(C(CO)N)O

Specifications

Melting Point 74°C
Color White-Yellow
Quantity 25 mg
Formula Weight 299.50
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material D-Sphingosine
TSCA No
Recommended Storage Freezer