missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Serine 98.5+%, TCI America™
$49.36 - $140.49
Chemical Identifiers
| CAS | 312-84-5 |
|---|---|
| Molecular Formula | C3H7NO3 |
| Molecular Weight (g/mol) | 105.093 |
| MDL Number | MFCD00004269 |
| InChI Key | MTCFGRXMJLQNBG-UWTATZPHSA-N |
| Synonym | d-serine, r-2-amino-3-hydroxypropanoic acid, 2r-2-amino-3-hydroxypropanoic acid, serine d-form, r-serine, serine, d, d-+-serine, h-d-ser-oh, d-2-amino-3-hydroxypropionic acid, d-serin |
| PubChem CID | 71077 |
| ChEBI | CHEBI:16523 |
| IUPAC Name | (2R)-2-amino-3-hydroxypropanoic acid |
| SMILES | C(C(C(=O)O)N)O |
Chemical Identifiers
| C3H7NO3 | |
| MFCD00004269 | |
| d-serine, r-2-amino-3-hydroxypropanoic acid, 2r-2-amino-3-hydroxypropanoic acid, serine d-form, r-serine, serine, d, d-+-serine, h-d-ser-oh, d-2-amino-3-hydroxypropionic acid, d-serin | |
| CHEBI:16523 | |
| C(C(C(=O)O)N)O |
Specifications
| 312-84-5 | |
| C3H7NO3 | |
| d-serine, r-2-amino-3-hydroxypropanoic acid, 2r-2-amino-3-hydroxypropanoic acid, serine d-form, r-serine, serine, d, d-+-serine, h-d-ser-oh, d-2-amino-3-hydroxypropionic acid, d-serin | |
| C(C(C(=O)O)N)O | |
| 105.093 | |
| CHEBI:16523 | |
| ≥98.5% (T) | |
| D-Serine |
| White | |
| MFCD00004269 | |
| MTCFGRXMJLQNBG-UWTATZPHSA-N | |
| (2R)-2-amino-3-hydroxypropanoic acid | |
| 71077 | |
| 105.09 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes