missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Serine, 99%

CAS: 312-84-5 | C3H7NO3 | 105.093 g/mol

Supplier:  Thermo Scientific Chemicals A1135314

 View more versions of this product

Catalog No. AAA1135314

Only null left
Explore available promotions

Description

Description

D-Serine is involved in the biosynthesis of purines, pyrimidines, and other amino acids. D-Serine also is an agonist of glycine site of the NMDA-type glutamate receptor. It also acts as Lacosamide intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Serine is involved in the biosynthesis of purines, pyrimidines, and other amino acids. D-Serine also is an agonist of glycine site of the NMDA-type glutamate receptor. It also acts as Lacosamide intermediate.

Solubility
Soluble in water (346 mg/ml at 20°C), and mineral acids. Insoluble in alcohol, ether, and benzene.

Notes
Store at 4°C. Store away from oxidizing agents. Keep container closed when not in use.
Specifications

Specifications

218°C to 220°C (decomposition)
312-84-5
C3H7NO3
1721403
d-serine, r-2-amino-3-hydroxypropanoic acid, 2r-2-amino-3-hydroxypropanoic acid, serine d-form, r-serine, serine, d, d-+-serine, h-d-ser-oh, d-2-amino-3-hydroxypropionic acid, d-serin
MTCFGRXMJLQNBG-UWTATZPHSA-N
(2R)-2-amino-3-hydroxypropanoic acid
105.093
CHEBI:16523
105.09
D-Serine
99%
MFCD00004269
14,8460
Soluble in water (346mg/ml at 20°C),and mineral acids. Insoluble in alcohol,ether,and benzene.
C(C(C(=O)O)N)O
25 g
71077
−13° (c=5 in 5N HCl)
99%

Chemical Identifiers

312-84-5
105.093
MTCFGRXMJLQNBG-UWTATZPHSA-N
71077
(2R)-2-amino-3-hydroxypropanoic acid
C3H7NO3
MFCD00004269
d-serine, r-2-amino-3-hydroxypropanoic acid, 2r-2-amino-3-hydroxypropanoic acid, serine d-form, r-serine, serine, d, d-+-serine, h-d-ser-oh, d-2-amino-3-hydroxypropionic acid, d-serin
CHEBI:16523
C(C(C(=O)O)N)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 206-229-4

RTECSNumber : VT8200000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only