missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D(-)-Salicin, 99+%
CAS: 138-52-3 | C13H18O7 | 286.27 g/mol
$93.41 - $925.81
Chemical Identifiers
| CAS | 138-52-3 |
|---|---|
| Molecular Formula | C13H18O7 |
| Molecular Weight (g/mol) | 286.27 |
| MDL Number | MFCD00006590 |
| InChI Key | NGFMICBWJRZIBI-UJPOAAIJSA-N |
| Synonym | salicin, salicoside, salicine, salicyl alcohol glucoside, d---salicin, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol, saligenin beta-d-glucopyranoside, 2-hydroxymethyl phenyl beta-d-glucopyranoside, d-salicin, saligenin-beta-d-glucopyranoside |
| PubChem CID | 439503 |
| ChEBI | CHEBI:17814 |
| IUPAC Name | (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol |
| SMILES | C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC132590050
|
Thermo Scientific Chemicals
132590050 |
5 g | Glass bottle |
Each for $93.41
|
|
||||
|
AC132590250
|
Thermo Scientific Chemicals
132590250 |
25 g | Glass bottle |
Each for $296.80
|
|
||||
|
AC132591000
|
Thermo Scientific Chemicals
132591000 |
100 g | Glass bottle |
Each for $925.81
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 138-52-3 | |
| 286.27 | |
| NGFMICBWJRZIBI-UJPOAAIJSA-N | |
| 439503 | |
| (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol |
| C13H18O7 | |
| MFCD00006590 | |
| salicin, salicoside, salicine, salicyl alcohol glucoside, d---salicin, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol, saligenin beta-d-glucopyranoside, 2-hydroxymethyl phenyl beta-d-glucopyranoside, d-salicin, saligenin-beta-d-glucopyranoside | |
| CHEBI:17814 | |
| C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O |
Specifications
| 138-52-3 | |
| White | |
| 99% min. (HPLC) | |
| C13H18O7 | |
| 5 g | |
| 15, 8460 | |
| -61.5 | |
| Solubility in water: 36g/L (15°C),250g/L (60°C). Other solubilities: insoluble in ether | |
| C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O | |
| 286.27 | |
| CHEBI:17814 | |
| 99+% | |
| D(-)-Salicin, 99+% |
| 197°C to 200°C | |
| Authentic | |
| Glass bottle | |
| MFCD00006590 | |
| 17, V,7, 113 | |
| −61.50° (20°C c=5,water) | |
| salicin, salicoside, salicine, salicyl alcohol glucoside, d---salicin, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol, saligenin beta-d-glucopyranoside, 2-hydroxymethyl phenyl beta-d-glucopyranoside, d-salicin, saligenin-beta-d-glucopyranoside | |
| NGFMICBWJRZIBI-UJPOAAIJSA-N | |
| (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol | |
| 439503 | |
| 286.27 | |
| Fine Crystalline Powder |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
GHS P Statement
Wear eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 205-331-6
RTECSNumber : LZ5901700
TSCA : TSCA
RUO – Research Use Only