missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Ribose-5-phosphate barium salt hexahydrate, 99%
CAS: 15673-79-7 | C5H9BaO8P | 365.42 g/mol
$218.90 - $628.45
Chemical Identifiers
| CAS | 15673-79-7 |
|---|---|
| Molecular Formula | C5H9BaO8P |
| Molecular Weight (g/mol) | 365.42 |
| MDL Number | MFCD00047534 |
| InChI Key | VNFDCDZWBYRPMC-QHEAFLAXSA-L |
| PubChem CID | 123134315 |
| SMILES | [Ba++].OC1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ66017MD
|
Thermo Scientific Chemicals
J66017MD |
250 mg |
Each for $218.90
|
|
|||||
|
AAJ6601703
|
Thermo Scientific Chemicals
J6601703 |
1 g |
Each for $628.45
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 15673-79-7 | |
| 365.42 | |
| VNFDCDZWBYRPMC-QHEAFLAXSA-L | |
| [Ba++].OC1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O |
| C5H9BaO8P | |
| MFCD00047534 | |
| 123134315 |
Specifications
| 15673-79-7 | |
| C5H9BaO8P | |
| 250 mg | |
| Hygroscopic | |
| +12° (c=1 in 0.2M HCl) | |
| barium(2+) [(2R,3S,4R)-3,4,5-trihydroxyoxolan-2-yl]methyl phosphate | |
| 123134315 | |
| 99% | |
| D-Ribose-5-phosphate barium salt hexahydrate |
| White to Yellow | |
| MFCD00047534 | |
| 3781061 | |
| VNFDCDZWBYRPMC-QHEAFLAXSA-L | |
| [Ba++].OC1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O | |
| 365.42 | |
| 473.51 | |
| Powder |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
EINECSNumber : 239-750-0
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only