missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(-)-Quinic acid, 98%
Description
D(-)-Quinic Acid is used in the synthesis of anti-influenza/anti-swine flu medication.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D(-)-Quinic Acid is used in the synthesis of anti-influenza/anti-swine flu medication.
Solubility
Soluble in water (400g/L), methanol and dimethyl sulfoxide.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
D(-)-Quinic Acid is used in the synthesis of anti-influenza/anti-swine flu medication.
Solubility
Soluble in water (400g/L), methanol and dimethyl sulfoxide.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 162°C to 167°C |
| Odor | Odorless |
| Quantity | 5 g |
| Beilstein | 2212412 |
| Merck Index | 14,8059 |
| Solubility Information | Soluble in water (400g/L),methanol and dimethyl sulfoxide. |
| Optical Rotation | −44° (c=20 in Water) |
| Formula Weight | 192.17 |
| Percent Purity | 98% |
| Chemical Name or Material | D-(-)-Quinic acid |
Safety and Handling
| Hazard Category | H319 |
| Hazard Statement | GHS H Statement H315-H319-H317-H335 Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. |
| Precautionary Statement | P264b-P280i-P305+P351+P338 |
| EINECSNumber | 201-072-8 |
| RTECSNumber | GU8650000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only