missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Quinic acid, 98%

Catalog No. AAL1523806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1523806 5 g
AAL1523814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1523806 Supplier Thermo Scientific Chemicals Supplier No. L1523806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 77-95-2 | C7H12O6 | 192.17 g/mol

D(-)-Quinic Acid is used in the synthesis of anti-influenza/anti-swine flu medication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D(-)-Quinic Acid is used in the synthesis of anti-influenza/anti-swine flu medication.

Solubility
Soluble in water (400g/L), methanol and dimethyl sulfoxide.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 77-95-2
Molecular Formula C7H12O6
Molecular Weight (g/mol) 192.17
MDL Number MFCD00003864
InChI Key AAWZDTNXLSGCEK-UHFFFAOYNA-N
Synonym quinic acid, d---quinic acid, --quinic acid, chinic acid, kinic acid, chinasaure, quinate, l-quinic acid, d-quinic acid, unii-058c04bgyi
PubChem CID 6508
IUPAC Name (3R,5R)-1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid
SMILES OC1CC(O)(CC(O)C1O)C(O)=O

Specifications

Melting Point 162°C to 167°C
Odor Odorless
Quantity 5 g
Beilstein 2212412
Merck Index 14,8059
Solubility Information Soluble in water (400g/L),methanol and dimethyl sulfoxide.
Optical Rotation −44° (c=20 in Water)
Formula Weight 192.17
Percent Purity 98%
Chemical Name or Material D-(-)-Quinic acid
Hazard Category H319
Hazard Statement GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 201-072-8
RTECSNumber GU8650000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only