missing translation for 'onlineSavingsMsg'
Learn More

D-Psicose 99.0+%, TCI America™

Catalog No. p-7135217
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1699100MG 100 mg
P16991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1699100MG Supplier TCI America Supplier No. P1699100MG

Chemical Identifiers

CAS 551-68-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00083478
InChI Key BJHIKXHVCXFQLS-PUFIMZNGSA-N
Synonym erythrohexulose, 3r,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, pseudofructose, unii-qcc18lng3e, d-ribo-hex-2-ulose, qcc18lng3e, dl-psicose, ribo-2-hexulose, d-ribo-2-ketohexose, keto-d-psicose
PubChem CID 90008
ChEBI CHEBI:27605
IUPAC Name (3R,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O

Specifications

Melting Point 109°C
Color White
Quantity 100 mg
Formula Weight 180.16
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material D-Psicose