missing translation for 'onlineSavingsMsg'
Learn More

D-Psicose, 98%

Catalog No. p-6926034
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456270010 1 g
AC456270050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC456270010 Supplier Thermo Scientific Chemicals Supplier No. 456270010
Only null left

CAS: 551-68-8 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 551-68-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00083478
InChI Key BJHIKXHVCXFQLS-PUFIMZNGSA-N
Synonym erythrohexulose, 3r,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, pseudofructose, unii-qcc18lng3e, d-ribo-hex-2-ulose, qcc18lng3e, dl-psicose, ribo-2-hexulose, d-ribo-2-ketohexose, keto-d-psicose
PubChem CID 90008
ChEBI CHEBI:27605
IUPAC Name (3R,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O

Specifications

Melting Point 109.0°C
Color White
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, insoluble in acetone
Formula Weight 180.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material D-Psicose
EINECSNumber 208-999-7

RUO – Research Use Only