missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Prolinol 99.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 76°C |
| Quantity | 5 g |
| Formula Weight | 101.15 |
| Percent Purity | ≥99.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Liquid |
| Chemical Name or Material | D-Prolinol |
Safety and Handling
| TSCA | No |