missing translation for 'onlineSavingsMsg'
Learn More

D(-)Prolinol, 99%

Catalog No. p-2691290
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC418310010 1 g Glass bottle
AC418310050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC418310010 Supplier Thermo Scientific Chemicals Supplier No. 418310010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 68832-13-3 | C5H11NO | 101.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68832-13-3
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00064321
InChI Key HVVNJUAVDAZWCB-RXMQYKEDSA-N
Synonym d-prolinol, r-pyrrolidin-2-ylmethanol, 2r-pyrrolidin-2-ylmethanol, r---2-pyrrolidinemethanol, r-prolinol, 2r-pyrrolidin-2-yl methanol, r-1-pyrrolidin-2-yl-methanol, 2-pyrrolidinemethanol, 2r, r---2-hydroxymethyl pyrrolidine
PubChem CID 2724541
ChEBI CHEBI:84258
IUPAC Name [(2R)-pyrrolidin-2-yl]methanol
SMILES C1CC(NC1)CO

Specifications

Infrared Spectrum Authentic
Color Amber to Colorless
Packaging Glass bottle
Refractive Index 1.4830 to 1.4870 (20°C, 589nm)
Specific Rotation −29° to −33° (20°C, 589nm) (c=1, toluene)
Density 1.02g/mL
Specific Rotation Condition −31° (20°C c=1,toluene)
Quantity 1 g
Physical Form Solution
Assay Percent Range 99%
Formula Weight 101.15
Percent Purity ≥98.5% (GC)
Chemical Name or Material D(-)Prolinol, 99%
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Recommended Storage Refrigerator +4°C

RUO – Research Use Only