missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Proline ethyl ester hydrochloride, 98%
CAS: 131477-20-8 | C7H14ClNO2 | 179.644 g/mol
$80.68 - $270.10
Chemical Identifiers
| CAS | 131477-20-8 |
|---|---|
| Molecular Formula | C7H14ClNO2 |
| Molecular Weight (g/mol) | 179.644 |
| MDL Number | MFCD00191003 |
| InChI Key | DUJGQVVONTYHLT-FYZOBXCZSA-N |
| Synonym | r-ethyl pyrrolidine-2-carboxylate hydrochloride, d-proline ethyl ester hydrochloride, h-d-pro-oet hcl, h-d-pro-oet hydrochloride, d-proline, ethyl ester, hydrochloride, ethyl 2r-pyrrolidine-2-carboxylate hydrochloride, h-d-pro-oet.hcl, h-d-pro-oet . hci, h-d-pro-oet?cl, d-proline ethyl ester hcl |
| PubChem CID | 56777374 |
| IUPAC Name | ethyl (2R)-pyrrolidine-2-carboxylate;hydrochloride |
| SMILES | CCOC(=O)C1CCCN1.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6491006
|
Thermo Scientific Chemicals
H6491006 |
5 g |
Each for $80.68
|
|
|||||
|
AAH6491014
|
Thermo Scientific Chemicals
H6491014 |
25 g |
Each for $270.10
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 131477-20-8 | |
| 179.644 | |
| DUJGQVVONTYHLT-FYZOBXCZSA-N | |
| 56777374 | |
| CCOC(=O)C1CCCN1.Cl |
| C7H14ClNO2 | |
| MFCD00191003 | |
| r-ethyl pyrrolidine-2-carboxylate hydrochloride, d-proline ethyl ester hydrochloride, h-d-pro-oet hcl, h-d-pro-oet hydrochloride, d-proline, ethyl ester, hydrochloride, ethyl 2r-pyrrolidine-2-carboxylate hydrochloride, h-d-pro-oet.hcl, h-d-pro-oet . hci, h-d-pro-oet?cl, d-proline ethyl ester hcl | |
| ethyl (2R)-pyrrolidine-2-carboxylate;hydrochloride |
Specifications
| 131477-20-8 | |
| C7H14ClNO2 | |
| 7170607 | |
| DUJGQVVONTYHLT-FYZOBXCZSA-N | |
| ethyl (2R)-pyrrolidine-2-carboxylate;hydrochloride | |
| 179.644 | |
| 179.64 | |
| Liquid |
| Yellow | |
| MFCD00191003 | |
| r-ethyl pyrrolidine-2-carboxylate hydrochloride, d-proline ethyl ester hydrochloride, h-d-pro-oet hcl, h-d-pro-oet hydrochloride, d-proline, ethyl ester, hydrochloride, ethyl 2r-pyrrolidine-2-carboxylate hydrochloride, h-d-pro-oet.hcl, h-d-pro-oet . hci, h-d-pro-oet?cl, d-proline ethyl ester hcl | |
| CCOC(=O)C1CCCN1.Cl | |
| 5 g | |
| 56777374 | |
| 98% | |
| D-Proline ethyl ester hydrochloride |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only