missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Proline, 99+%

CAS: 344-25-2 | C5H9NO2 | 115.132 g/mol

$81.45 - $81.45

Chemical Identifiers

CAS 344-25-2
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00064317
InChI Key ONIBWKKTOPOVIA-SCSAIBSYSA-N
Synonym d-proline, 2r-pyrrolidine-2-carboxylic acid, r-pyrrolidine-2-carboxylic acid, h-d-pro-oh, d-pro, d-+-proline, d-prolin, proline, r-proline, d +-proline
PubChem CID 8988
ChEBI CHEBI:16313
IUPAC Name (2R)-pyrrolidine-2-carboxylic acid
SMILES C1CC(NC1)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC228660010
SDS View Documents
Thermo Scientific Chemicals
228660010
1 g Glass Bottle
Each for $81.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

344-25-2
115.132
ONIBWKKTOPOVIA-SCSAIBSYSA-N
8988
(2R)-pyrrolidine-2-carboxylic acid
C5H9NO2
MFCD00064317
d-proline, 2r-pyrrolidine-2-carboxylic acid, r-pyrrolidine-2-carboxylic acid, h-d-pro-oh, d-pro, d-+-proline, d-prolin, proline, r-proline, d +-proline
CHEBI:16313
C1CC(NC1)C(=O)O

Specifications

344-25-2
Authentic
99+%
C5H9NO2
MFCD00064317
22, 2
0.1 % max.
ONIBWKKTOPOVIA-SCSAIBSYSA-N
(2R)-pyrrolidine-2-carboxylic acid
115.132
8988
115.13
Powder
Brown to White
1 % max. (105°C, 3 hrs)
0.5% max. (TLC)
Glass Bottle
+83.0° to +87.0° (20°C, 589nm) (c=4, water)
07,307; 09,393
d-proline, 2r-pyrrolidine-2-carboxylic acid, r-pyrrolidine-2-carboxylic acid, h-d-pro-oh, d-pro, d-+-proline, d-prolin, proline, r-proline, d +-proline
C1CC(NC1)C(=O)O
+84.75 (20.00°C c=4,H2O)
1 g
CHEBI:16313
99.0 to 101.0% (Total base)
D(+)-Proline
Videos
SDS
Documents

Documents

RUO – Research Use Only