missing translation for 'onlineSavingsMsg'
Learn More

D-Pipecolinic acid, 97%

CAS: 1723-00-8 | C6H11NO2 | 129.159 g/mol

Supplier:  Thermo Scientific Chemicals H6325522

 View more versions of this product

Catalog No. AAH6325522


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

D-Pipecolinic acid
277°C (decomposition)
MFCD00064346
81094
HXEACLLIILLPRG-RXMQYKEDSA-N
(2R)-piperidine-2-carboxylic acid
736316
129.16
Solid
1723-00-8
C6H11NO2
100 g
r-piperidine-2-carboxylic acid, d-pipecolic acid, d-pipecolinic acid, d +-pipecolinic acid, d-homoproline, 2r-piperidine-2-carboxylic acid, r-pipecolic acid, +-pipecolic acid, d-piperidine-2-carboxylic acid
C1CCNC(C1)C(=O)O
129.159
CHEBI:41582
97%

Chemical Identifiers

1723-00-8
129.159
HXEACLLIILLPRG-RXMQYKEDSA-N
736316
(2R)-piperidine-2-carboxylic acid
C6H11NO2
MFCD00064346
r-piperidine-2-carboxylic acid, d-pipecolic acid, d-pipecolinic acid, d +-pipecolinic acid, d-homoproline, 2r-piperidine-2-carboxylic acid, r-pipecolic acid, +-pipecolic acid, d-piperidine-2-carboxylic acid
CHEBI:41582
C1CCNC(C1)C(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 217-024-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only