missing translation for 'onlineSavingsMsg'
Learn More

D-Pipecolic Acid 98.0+%, TCI America™

Catalog No. p-7135277
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18305G 5 g
P183025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18305G Supplier TCI America Supplier No. P18305G
Only null left

Chemical Identifiers

CAS 1723-00-8
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00064346
InChI Key HXEACLLIILLPRG-RXMQYKEDSA-N
Synonym r-piperidine-2-carboxylic acid, d-pipecolic acid, d-pipecolinic acid, d +-pipecolinic acid, d-homoproline, 2r-piperidine-2-carboxylic acid, r-pipecolic acid, +-pipecolic acid, d-piperidine-2-carboxylic acid
PubChem CID 736316
ChEBI CHEBI:41582
IUPAC Name (2R)-piperidine-2-carboxylic acid
SMILES C1CCNC(C1)C(=O)O

Specifications

Melting Point 270°C
Color White-Yellow
Quantity 5 g
Formula Weight 129.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-Pipecolic Acid