missing translation for 'onlineSavingsMsg'
Learn More

D-Phenylalanine 98.0+%, TCI America™

Catalog No. p-7134440
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P01355G 5 g
P013525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P01355G Supplier TCI America Supplier No. P01355G

Chemical Identifiers

CAS 673-06-3
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00004270
InChI Key COLNVLDHVKWLRT-MRVPVSSYSA-N
Synonym d-phenylalanine, h-d-phe-oh, 2r-2-amino-3-phenylpropanoic acid, d-phe, sabiden, phenylalanine d-form, alanine, phenyl-, d, d-diphenylalanine, r-2-amino-3-phenylpropanoic acid, d-alpha-amino-beta-phenylpropionic acid
PubChem CID 71567
ChEBI CHEBI:16998
IUPAC Name (2R)-2-amino-3-phenylpropanoic acid
SMILES N[C@H](CC1=CC=CC=C1)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 165.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-Phenylalanine
EINECSNumber (9)-1282
RTECSNumber AY7533000
TSCA Yes