missing translation for 'onlineSavingsMsg'
Learn More

D-Phenylalanine Methyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. p-7135226
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17255G 5 g
P172525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17255G Supplier TCI America Supplier No. P17255G

Chemical Identifiers

CAS 13033-84-6
Molecular Formula C10H14ClNO2
Molecular Weight (g/mol) 215.68
MDL Number MFCD00066112
InChI Key SWVMLNPDTIFDDY-SBSPUUFOSA-N
Synonym d-phenylalanine methyl ester hydrochloride, h-d-phe-ome.hcl, d-phenylalanine methyl ester hcl, h-d-phe-ome hcl, r-methyl 2-amino-3-phenylpropanoate hydrochloride, d-phenylalanine, methyl ester, hydrochloride, methyl d-phenylalaninate hydrochloride, r-phenylalanine methyl ester hydrochloride, methyl 2r-2-amino-3-phenylpropanoate hydrochloride
PubChem CID 11481330
IUPAC Name methyl (2R)-2-amino-3-phenylpropanoate hydrochloride
SMILES Cl.COC(=O)[C@H](N)CC1=CC=CC=C1

Specifications

Melting Point 160°C
Color White
Quantity 5 g
Formula Weight 215.68
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material D-Phenylalanine Methyl Ester Hydrochloride