missing translation for 'onlineSavingsMsg'
Learn More

D-Phenylalanine methyl ester hydrochloride, 98%

Catalog No. AAH2592006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2592006 5 g
AAH2592014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2592006 Supplier Thermo Scientific Chemicals Supplier No. H2592006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13033-84-6 | C10H14ClNO2 | 215.68 g/mol

Benzhydrylidene Aza-Glycinyl-D-Phenylalanine Methyl Ester was synthesized from D-phenylalanine methyl ester hydrochloride. Used in the conversion of amino acids into n-menthyloxycarbonyl alkyl ester derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzhydrylidene Aza-Glycinyl-D-Phenylalanine Methyl Ester was synthesized from D-phenylalanine methyl ester hydrochloride. Used in the conversion of amino acids into n-menthyloxycarbonyl alkyl ester derivatives.

Solubility
Soluble in Ethanol and Methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.

Chemical Identifiers

CAS 13033-84-6
Molecular Formula C10H14ClNO2
Molecular Weight (g/mol) 215.68
MDL Number MFCD00066112
InChI Key SWVMLNPDTIFDDY-SBSPUUFOSA-N
Synonym d-phenylalanine methyl ester hydrochloride, h-d-phe-ome.hcl, d-phenylalanine methyl ester hcl, h-d-phe-ome hcl, r-methyl 2-amino-3-phenylpropanoate hydrochloride, d-phenylalanine, methyl ester, hydrochloride, methyl d-phenylalaninate hydrochloride, r-phenylalanine methyl ester hydrochloride, methyl 2r-2-amino-3-phenylpropanoate hydrochloride
PubChem CID 11481330
IUPAC Name methyl (2R)-2-amino-3-phenylpropanoate;hydrochloride
SMILES Cl.COC(=O)[C@H](N)CC1=CC=CC=C1

Specifications

Melting Point 158°C to 162°C
Solubility Information Soluble in Ethanol and Methanol.
Quantity 5 g
Formula Weight 215.68
Percent Purity 99%
Chemical Name or Material D-Phenylalanine methyl ester hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only