missing translation for 'onlineSavingsMsg'
Learn More

D-Phenylalanine methyl ester hydrochloride, 98%

CAS: 13033-84-6 | C10H14ClNO2 | 215.68 g/mol

$89.33 - $89.33

Chemical Identifiers

CAS 13033-84-6
Molecular Formula C10H14ClNO2
Molecular Weight (g/mol) 215.68
MDL Number MFCD00066112
InChI Key SWVMLNPDTIFDDY-SBSPUUFOSA-N
Synonym d-phenylalanine methyl ester hydrochloride, h-d-phe-ome.hcl, d-phenylalanine methyl ester hcl, h-d-phe-ome hcl, r-methyl 2-amino-3-phenylpropanoate hydrochloride, d-phenylalanine, methyl ester, hydrochloride, methyl d-phenylalaninate hydrochloride, r-phenylalanine methyl ester hydrochloride, methyl 2r-2-amino-3-phenylpropanoate hydrochloride
PubChem CID 11481330
IUPAC Name methyl (2R)-2-amino-3-phenylpropanoate;hydrochloride
SMILES Cl.COC(=O)[C@H](N)CC1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC442620050
SDS View Documents
Thermo Scientific Chemicals
442620050
5 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13033-84-6
215.68
SWVMLNPDTIFDDY-SBSPUUFOSA-N
11481330
Cl.COC(=O)[C@H](N)CC1=CC=CC=C1
C10H14ClNO2
MFCD00066112
d-phenylalanine methyl ester hydrochloride, h-d-phe-ome.hcl, d-phenylalanine methyl ester hcl, h-d-phe-ome hcl, r-methyl 2-amino-3-phenylpropanoate hydrochloride, d-phenylalanine, methyl ester, hydrochloride, methyl d-phenylalaninate hydrochloride, r-phenylalanine methyl ester hydrochloride, methyl 2r-2-amino-3-phenylpropanoate hydrochloride
methyl (2R)-2-amino-3-phenylpropanoate;hydrochloride

Specifications

13033-84-6
White
97.5% min. (HPLC)
MFCD00066112
-35.76
SWVMLNPDTIFDDY-SBSPUUFOSA-N
methyl (2R)-2-amino-3-phenylpropanoate;hydrochloride
11481330
98%
D-Phenylalanine methyl ester hydrochloride
157°C to 159°C
Authentic
C10H14ClNO2
5 g
d-phenylalanine methyl ester hydrochloride, h-d-phe-ome.hcl, d-phenylalanine methyl ester hcl, h-d-phe-ome hcl, r-methyl 2-amino-3-phenylpropanoate hydrochloride, d-phenylalanine, methyl ester, hydrochloride, methyl d-phenylalaninate hydrochloride, r-phenylalanine methyl ester hydrochloride, methyl 2r-2-amino-3-phenylpropanoate hydrochloride
Cl.COC(=O)[C@H](N)CC1=CC=CC=C1
215.68
215.68
Crystalline Powder or Powder
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only