missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Phenylalanine, 99%

CAS: 673-06-3 | C9H11NO2 | 165.19 g/mol

Supplier:  Thermo Scientific Chemicals A1057214

 View more versions of this product

Catalog No. AAA1057214


Only null left

Description

Description

It is an essential amino acid with anti-depressant and analgesic activities. pharmaceutical intermediates, mainly used to treat, prevent osteoporosis, cardiovascular, diabetes, arterioscleros.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an essential amino acid with anti-depressant and analgesic activities. pharmaceutical intermediates, mainly used to treat, prevent osteoporosis, cardiovascular, diabetes, arterioscleros.

Solubility
Soluble in water (30 mg/ml). Slightly soluble in methanol, ethanol, DMSO.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications

Specifications

∽273°C to 276°C (decomposition)
673-06-3
MFCD00004270
14,7271
Soluble in water (30mg/ml). Slightly soluble in methanol,ethanol,DMSO.
N[C@H](CC1=CC=CC=C1)C(O)=O
25 g
71567
+34° (c=2 in water)
99%
D-Phenylalanine
C9H11NO2
2804068
d-phenylalanine, h-d-phe-oh, 2r-2-amino-3-phenylpropanoic acid, d-phe, sabiden, phenylalanine d-form, alanine, phenyl-, d, d-diphenylalanine, r-2-amino-3-phenylpropanoic acid, d-alpha-amino-beta-phenylpropionic acid
COLNVLDHVKWLRT-MRVPVSSYSA-N
(2R)-2-amino-3-phenylpropanoic acid
165.19
CHEBI:16998
165.19

Chemical Identifiers

673-06-3
165.19
COLNVLDHVKWLRT-MRVPVSSYSA-N
71567
(2R)-2-amino-3-phenylpropanoic acid
C9H11NO2
MFCD00004270
d-phenylalanine, h-d-phe-oh, 2r-2-amino-3-phenylpropanoic acid, d-phe, sabiden, phenylalanine d-form, alanine, phenyl-, d, d-diphenylalanine, r-2-amino-3-phenylpropanoic acid, d-alpha-amino-beta-phenylpropionic acid
CHEBI:16998
N[C@H](CC1=CC=CC=C1)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-603-5

RTECSNumber : AY7533000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only