missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Phenylalanine, 99%

Catalog No. AAA1057222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1057222 100 g
AAA1057206 5 g
AAA1057214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1057222 Supplier Thermo Scientific Chemicals Supplier No. A1057222

CAS: 673-06-3 | C9H11NO2 | 165.19 g/mol

It is an essential amino acid with anti-depressant and analgesic activities. pharmaceutical intermediates, mainly used to treat, prevent osteoporosis, cardiovascular, diabetes, arterioscleros.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an essential amino acid with anti-depressant and analgesic activities. pharmaceutical intermediates, mainly used to treat, prevent osteoporosis, cardiovascular, diabetes, arterioscleros.

Solubility
Soluble in water (30 mg/ml). Slightly soluble in methanol, ethanol, DMSO.

Notes
Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 673-06-3
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00004270
InChI Key COLNVLDHVKWLRT-MRVPVSSYSA-N
Synonym d-phenylalanine, h-d-phe-oh, 2r-2-amino-3-phenylpropanoic acid, d-phe, sabiden, phenylalanine d-form, alanine, phenyl-, d, d-diphenylalanine, r-2-amino-3-phenylpropanoic acid, d-alpha-amino-beta-phenylpropionic acid
PubChem CID 71567
ChEBI CHEBI:16998
IUPAC Name (2R)-2-amino-3-phenylpropanoic acid
SMILES N[C@H](CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point ∽273°C to 276°C (decomposition)
Beilstein 2804068
Merck Index 14,7271
Solubility Information Soluble in water (30mg/ml). Slightly soluble in methanol,ethanol,DMSO.
Quantity 100 g
Optical Rotation +34° (c=2 in water)
Formula Weight 165.19
Percent Purity 99%
Chemical Name or Material D-Phenylalanine
EINECSNumber 211-603-5
RTECSNumber AY7533000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only