missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Phenylalanine, 99+%

Catalog No. AC172041000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC172041000 100 g Glass Bottle
AC172040250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC172041000 Supplier Thermo Scientific Chemicals Supplier No. 172041000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 673-06-3 | C9H11NO2 | 165.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 673-06-3
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00004270
InChI Key COLNVLDHVKWLRT-MRVPVSSYSA-N
Synonym d-phenylalanine, h-d-phe-oh, 2r-2-amino-3-phenylpropanoic acid, d-phe, sabiden, phenylalanine d-form, alanine, phenyl-, d, d-diphenylalanine, r-2-amino-3-phenylpropanoic acid, d-alpha-amino-beta-phenylpropionic acid
PubChem CID 71567
ChEBI CHEBI:16998
IUPAC Name (2R)-2-amino-3-phenylpropanoic acid
SMILES N[C@H](CC1=CC=CC=C1)C(O)=O

Specifications

Arsenic (As) 3ppm max.
Color White
Heavy Metals (as Pb) 20ppm max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Loss on Drying 0.5% max.
Packaging Glass Bottle
Linear Formula C6H5CH2CH(NH2)CO2H
Beilstein 14, 494
Specific Rotation +32° to +35° (20°C, 589nm) (c=2, water)
Merck Index 15, 7382
Sulfated Ash 0.1% max.
Specific Rotation Condition +33.50° (20°C c=2,H2O)
Quantity 100 g
Formula Weight 165.19
Percent Purity ≥99.0%
Physical Form Powder
Chemical Name or Material D-Phenylalanine
Show More Show Less

RUO – Research Use Only