missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Penicillamine 98.0+%, TCI America™
$266.56 - $797.81
Chemical Identifiers
| CAS | 52-67-5 |
|---|---|
| Molecular Formula | C5H11NO2S |
| Molecular Weight (g/mol) | 149.208 |
| MDL Number | MFCD00064302 |
| InChI Key | VVNCNSJFMMFHPL-VKHMYHEASA-N |
| Synonym | d-penicillamine, penicillamine, cuprimine, d---penicillamine, depen, 3-mercapto-d-valine, cuprenil, d-penamine, --penicillamine, d-mercaptovaline |
| PubChem CID | 5852 |
| ChEBI | CHEBI:7959 |
| IUPAC Name | (2S)-2-amino-3-methyl-3-sulfanylbutanoic acid |
| SMILES | CC(C)(C(C(=O)O)N)S |
Chemical Identifiers
| 52-67-5 | |
| 149.208 | |
| VVNCNSJFMMFHPL-VKHMYHEASA-N | |
| 5852 | |
| (2S)-2-amino-3-methyl-3-sulfanylbutanoic acid |
| C5H11NO2S | |
| MFCD00064302 | |
| d-penicillamine, penicillamine, cuprimine, d---penicillamine, depen, 3-mercapto-d-valine, cuprenil, d-penamine, --penicillamine, d-mercaptovaline | |
| CHEBI:7959 | |
| CC(C)(C(C(=O)O)N)S |
Specifications
| 52-67-5 | |
| C5H11NO2S | |
| 5 g | |
| VVNCNSJFMMFHPL-VKHMYHEASA-N | |
| (2S)-2-amino-3-methyl-3-sulfanylbutanoic acid | |
| 5852 | |
| 149.21 | |
| Crystalline Powder |
| White | |
| MFCD00064302 | |
| d-penicillamine, penicillamine, cuprimine, d---penicillamine, depen, 3-mercapto-d-valine, cuprenil, d-penamine, --penicillamine, d-mercaptovaline | |
| CC(C)(C(C(=O)O)N)S | |
| 149.208 | |
| CHEBI:7959 | |
| ≥98.0% (T) | |
| D-Penicillamine |
Safety and Handling
RTECSNumber : YV9425000
TSCA : No