missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Penicillamine, 98%

Catalog No. AAA1144606
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1144606 5 g
AAA1144614 25 g
AAA1144622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1144606 Supplier Thermo Scientific Chemicals Supplier No. A1144606

CAS: 52-67-5 | C5H11NO2S | 149.208 g/mol

It is used mainly as chelating agent in heavy metal poisoning as in lead, mercury and copper poisoning. It is used in Wilson's disease. It  is used in pharmacy as chelating agent and as immunosuppressant for treating rheumatoid arthritis by reducing the number of T cells, inhibiting the macrophages, by reducing the activity of Interleukin 1 and rheumatoid factor and in preventing crosslinking of the collagen. It acts as an exogenous NOS modulator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
It is freely soluble in water, slightly soluble in alcohol, insoluble in chloroform, ether, acetone, benzene, and in carbon tetrachloride.

Notes
Stable under normal conditions. Incompatabile to strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C.

Chemical Identifiers

CAS 52-67-5
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.208
MDL Number MFCD00064302
InChI Key VVNCNSJFMMFHPL-VKHMYHEASA-N
Synonym d-penicillamine, penicillamine, cuprimine, d---penicillamine, depen, 3-mercapto-d-valine, cuprenil, d-penamine, --penicillamine, d-mercaptovaline
PubChem CID 5852
ChEBI CHEBI:7959
IUPAC Name (2S)-2-amino-3-methyl-3-sulfanylbutanoic acid
SMILES CC(C)(C(C(=O)O)N)S

Specifications

Melting Point 202°C to 204°C (decomposition)
pH 4.5 to 5.5
Odor Unpleasant
Quantity 5 g
Beilstein 1722375
Merck Index 14,7088
Solubility Information It is freely soluble in water,slightly soluble in alcohol,insoluble in chloroform,ether,acetone,benzene,and in carbon tetrachloride.
Optical Rotation −63° (c=1 in 1N NaOH)
Formula Weight 149.21
Percent Purity 99%
Chemical Name or Material D- (-)-Penicillamine
Show More Show Less
Hazard Category H315-H319-H334
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P280-P285-P302+P352-P304+P341-P305+P351+P338-P332+P313-P342+P311-P362-P501c
EINECSNumber 200-148-8
RTECSNumber YV9425000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only