missing translation for 'onlineSavingsMsg'
Learn More

D-Pantothenic Acid, Calcium Salt, MP Biomedicals™

Catalog No. ICN10122890
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10122890 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10122890 Supplier MP Biomedicals Inc Supplier No. 0210122890

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

D-Pantothenic acid is an essential vitamin (except in horses, ruminants).

Chemical Identifiers

CAS 137-08-6
Molecular Formula C18H32CaN2O10
Molecular Weight (g/mol) 476.54
MDL Number MFCD00002766
InChI Key FAPWYRCQGJNNSJ-DXHDTSSINA-L
Synonym calcium pantothenate, d-pantothenic acid hemicalcium salt, d-pantothenic acid, calcium salt, pantothenic acid, calcium salt, d, r-n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-beta-alanine calcium salt, beta-alanine, n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-, calcium salt, r
PubChem CID 131847364
IUPAC Name calcium bis(3-[(2R)-2,4-dihydroxy-3,3-dimethylbutanamido]propanoate)
SMILES [Ca++].CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O.CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 500 g
Solubility Information Soluble in water (1g/2.8mL; pH of a 5% aqueous solution is 7.2-8.0; CO2 free water: 8.7), glycerol; slightly soluble in ethanol or acetone.
Formula Weight 238.3
Grade USP
Recommended Storage Room Temperature (15–30°C)