missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Pantolactone, 99%

Catalog No. AC218140050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC218140050 5 g Glass bottle
AC218140250 25 g Glass bottle
AC218141000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC218140050 Supplier Thermo Scientific Chemicals Supplier No. 218140050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 599-04-2 | C6H10O3 | 130.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 599-04-2
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00005392
InChI Key SERHXTVXHNVDKA-BYPYZUCNSA-N
Synonym d---pantolactone, r-pantolactone, d-pantolactone, r-pantoyl lactone, d---pantoyl lactone, pantothenic lactone, r-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one, unii-j288d7o0js, 2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3r, r---pantolactone
PubChem CID 439368
ChEBI CHEBI:16719
IUPAC Name (3R)-3-hydroxy-4,4-dimethyloxolan-2-one
SMILES CC1(COC(=O)C1O)C

Specifications

Melting Point 88.0°C to 92.0°C
Color White
Boiling Point 120.0°C to 122.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 18, V,1, 22
Fieser 16,269; 17,270
Merck Index 15, 7116
Specific Rotation Condition −50.50° (25°C c=2,H2O)
Specific Rotation − 50.50
Solubility Information Solubility in water: soluble.
Formula Weight 130.14
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material D(-)-Pantolactone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-963-3

RUO – Research Use Only