Learn More
D-(-)-Pantolactone, 99%
CAS: 599-04-2 | C6H10O3 | 130.143 g/mol
Supplier: Thermo Scientific Chemicals L1598314
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| D-(-)-Pantolactone | |
| 89°C to 91°C | |
| C6H10O3 | |
| 25 g | |
| Hygroscopic | |
| d---pantolactone, r-pantolactone, d-pantolactone, r-pantoyl lactone, d---pantoyl lactone, pantothenic lactone, r-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one, unii-j288d7o0js, 2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3r, r---pantolactone | |
| −51° (c=3 in Water) | |
| (3R)-3-hydroxy-4,4-dimethyloxolan-2-one | |
| 439368 | |
| 130.14 |
| 599-04-2 | |
| 120°C to 122°C (15 mmHg) | |
| MFCD00005392 | |
| 80957 | |
| 14,7013 | |
| SERHXTVXHNVDKA-BYPYZUCNSA-N | |
| CC1(COC(=O)C1O)C | |
| 130.143 | |
| CHEBI:16719 | |
| 99% |
Chemical Identifiers
| 599-04-2 | |
| 130.143 | |
| SERHXTVXHNVDKA-BYPYZUCNSA-N | |
| 439368 | |
| (3R)-3-hydroxy-4,4-dimethyloxolan-2-one |
| C6H10O3 | |
| MFCD00005392 | |
| d---pantolactone, r-pantolactone, d-pantolactone, r-pantoyl lactone, d---pantoyl lactone, pantothenic lactone, r-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one, unii-j288d7o0js, 2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3r, r---pantolactone | |
| CHEBI:16719 | |
| CC1(COC(=O)C1O)C |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-963-3
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only