missing translation for 'onlineSavingsMsg'
Learn More

D-Panthenol 98.0+%, TCI America™

Catalog No. P069225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P069225G 25 g
P0692500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P069225G Supplier TCI America Supplier No. P069225G

Chemical Identifiers

CAS 81-13-0
Molecular Formula C9H19NO4
Molecular Weight (g/mol) 205.25
MDL Number MFCD00065006
InChI Key SNPLKNRPJHDVJA-ZETCQYMHSA-N
Synonym dexpanthenol, d-panthenol, pantothenol, bepanthen, d-pantothenyl alcohol, ilopan, +-panthenol, bepanthene, panthoderm, thenalton
PubChem CID 131204
ChEBI CHEBI:27373
IUPAC Name (2R)-2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
SMILES CC(C)(CO)[C@@H](O)C(=O)NCCCO

Specifications

Color Colorless
Quantity 25 g
Formula Weight 205.25
Percent Purity ≥98.0% (T)
Physical Form Starch Syrup
Chemical Name or Material D-Panthenol
EINECSNumber (9)-0998
RTECSNumber ES4316000
TSCA Yes